Learn More
Nabertherm High-Temperature Furnace, 1600 HT Series
Developed for applications requiring fast heating up ramps. Gas-fired high-temperature furnaces are preferred if inflammable gases are produced in large amounts during the process. Insulated with highly heat-resistant and long-life lightweight refractory brick insulation or fiber materials
29894.40€ - 89485.71€
| Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
| 15968290 |
Nabertherm
HT0426QNA |
40 L |
29894.40€ Each |
|
15998290 |
Nabertherm
HT0626QNA |
64 L |
37435.89€ Each |
|
15918300 |
Nabertherm
HT1226QNA |
128 L |
52802.74€ Each |
|
15938300 |
Nabertherm
HT1626QNA |
160 L |
45335.31€ Each |
|
15958300 |
Nabertherm
HT2726QNA |
276 L |
72279.77€ Each |
|
15978300 |
Nabertherm
HT4526QNA |
450 L |
89485.71€ Each |
|
Description
- Maximum Temperature: 1600 °C
- Powerful, sturdy high-speed burners with pulse control and special flame guidance in the furnace chamber provide for good temperature uniformity
- Operation with natural gas, propane or liquified gas
- Fully automatic PLC control of the temperature, including monitoring of the burner function
- Gas fittings according to DVGW (German Technical and Scientific Association for Gas and Water) with flame monitoring and safety valve
- Reduction-resistant fiber insulation with low heat storage provides for short heating and cooling times
- Dual shell housing provides for low outside temperatures
- Exhaust hood with fittings for further discharge of the exhaust gases
- Defined application within the constraints of the operating instructions
- NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
- PLC control with touch panel as user interface