Learn More
Nabertherm High-Temperature Furnace, 1600 HT Series
Developed for applications requiring fast heating up ramps. Gas-fired high-temperature furnaces are preferred if inflammable gases are produced in large amounts during the process. Insulated with highly heat-resistant and long-life lightweight refractory brick insulation or fiber materials
29894.40€ - 89485.71€
Products 6
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15968290 View Documents Nabertherm
HT0426QNA
40 L 29894.40€
Each View alternative products
Save Up to 
»
please call +351214253350
Add to basket
15998290 View Documents Nabertherm
HT0626QNA
64 L 37435.89€
Each View alternative products
Save Up to 
»
please call +351214253350
Add to basket
15918300 View Documents Nabertherm
HT1226QNA
128 L 52802.74€
Each View alternative products
Save Up to 
»
please call +351214253350
Add to basket
15938300 View Documents Nabertherm
HT1626QNA
160 L 45335.31€
Each View alternative products
Save Up to 
»
please call +351214253350
Add to basket
15958300 View Documents Nabertherm
HT2726QNA
276 L 72279.77€
Each View alternative products
Save Up to 
»
please call +351214253350
Add to basket
15978300 View Documents Nabertherm
HT4526QNA
450 L 89485.71€
Each View alternative products
Save Up to 
»
please call +351214253350
Add to basket
Description
Description
  • Maximum Temperature: 1600 °C
  • Powerful, sturdy high-speed burners with pulse control and special flame guidance in the furnace chamber provide for good temperature uniformity
  • Operation with natural gas, propane or liquified gas
  • Fully automatic PLC control of the temperature, including monitoring of the burner function
  • Gas fittings according to DVGW (German Technical and Scientific Association for Gas and Water) with flame monitoring and safety valve
  • Reduction-resistant fiber insulation with low heat storage provides for short heating and cooling times
  • Dual shell housing provides for low outside temperatures
  • Exhaust hood with fittings for further discharge of the exhaust gases
  • Defined application within the constraints of the operating instructions
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
  • PLC control with touch panel as user interface