Learn More
Nabertherm High-Temperature Furnace, 1700 HFL Series
Lined with robust refractory insulation. Compared with the fiber-insulated models of the HT product line, these furnaces are recommended when high charge weights have to be sintered.
25006.63€ - 68601.60€
| Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
| 15988280 |
Nabertherm
HF0327QNA |
16 L |
25006.63€ Each |
|
15908290 |
Nabertherm
HF0427QNA |
40 L |
40484.57€ Each |
|
15928290 |
Nabertherm
HF0627QNA |
64 L |
53666.74€ Each |
|
15948290 |
Nabertherm
HF1627QNA |
160 L |
68601.60€ Each |
|
Description
- Maximum Temperature: 1700 °C
- Robust refractory brick insulation and special backing insulation
- Furnace floor made of lightweight refractory bricks accommodates high charge weights
- Defined application within the constraints of the operating instructions
- NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive