Learn More
Nabertherm High-Temperature Furnace, 1700 HFL Series
Lined with robust refractory insulation. Compared with the fiber-insulated models of the HT product line, these furnaces are recommended when high charge weights have to be sintered.
25006.63€ - 68601.60€
Products 4
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15988280 View Documents Nabertherm
HF0327QNA
16 L 25006.63€
Each View alternative products
Save Up to 
»
please call +351214253350
Add to basket
15908290 View Documents Nabertherm
HF0427QNA
40 L 40484.57€
Each View alternative products
Save Up to 
»
please call +351214253350
Add to basket
15928290 View Documents Nabertherm
HF0627QNA
64 L 53666.74€
Each View alternative products
Save Up to 
»
please call +351214253350
Add to basket
15948290 View Documents Nabertherm
HF1627QNA
160 L 68601.60€
Each View alternative products
Save Up to 
»
please call +351214253350
Ajouter au panier
Description
Description
  • Maximum Temperature: 1700 °C
  • Robust refractory brick insulation and special backing insulation
  • Furnace floor made of lightweight refractory bricks accommodates high charge weights
  • Defined application within the constraints of the operating instructions
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive