Learn More
Nabertherm High-Temperature Furnace, 1750 HT Series
Whether for quartz glass or glass ceramics, for sintering CIM components or for other processes up to a maximum temperature of 1750 °C, these furnaces afford the optimal solution for the sintering process.
18653.14€ - 109555.20€
Products 7
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15958290 View Documents Nabertherm
HT0327QNA
16 L 18653.14€
Each View alternative products
Save Up to 
»
please call +351214253350
Add to basket
15978290 View Documents Nabertherm
HT0427QNA
40 L 32227.20€
Each View alternative products
Save Up to 
»
please call +351214253350
Add to basket
15908300 View Documents Nabertherm
HT0627QNA
64 L 42730.97€
Each View alternative products
Save Up to 
»
please call +351214253350
Add to basket
15928300 View Documents Nabertherm
HT1227QNA
128 L 60739.20€
Each View alternative products
Save Up to 
»
please call +351214253350
Add to basket
15948300 View Documents Nabertherm
HT1627QNA
160 L 54617.14€
Each View alternative products
Save Up to 
»
please call +351214253350
Add to basket
15968300 View Documents Nabertherm
HT2727QNA
276 L 84128.91€
Each View alternative products
Save Up to 
»
please call +351214253350
Add to basket
15988300 View Documents Nabertherm
HT4527QNA
450 L 109555.20€
Each View alternative products
Save Up to 
»
please call +351214253350
Add to basket
Description
Description
  • Maximum Temperature: 1750 °C
  • Dual shell housing with fan cooling for low shell temperatures
  • Heating from both sides via molybdenum disilicide heating elements
  • High-quality fiber insulation backed by special insulation
  • Side insulation constructed with tongue and groove blocks provides for low heat loss to the outside
  • Long-life roof insulation with special suspension
  • Chain-guided parallel swivel door for defined opening and closing of the door
  • Two-door design (front/back) for high-temperature furnaces > HT 276/..
  • Labyrinth sealing ensures the least possible temperature loss in the door area
  • Reinforced floor as protection for fiber insulation as standard from models HT 16/16 upwards
  • Exhaust air opening in the furnace roof
  • Heating elements switched via thyristors
  • Over-temperature limiter with adjustable cutout temperature for thermal protection class 2 in accordance with EN 60519-2 as temperature limiter to protect the furnace and load
  • Defined application within the constraints of the operating instructions
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive