Learn More
Nabertherm High-Temperature Furnace, 1750 HT Series
Whether for quartz glass or glass ceramics, for sintering CIM components or for other processes up to a maximum temperature of 1750 °C, these furnaces afford the optimal solution for the sintering process.
18653.14€ - 109555.20€
| Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
| 15958290 |
Nabertherm
HT0327QNA |
16 L |
18653.14€ Each |
|
15978290 |
Nabertherm
HT0427QNA |
40 L |
32227.20€ Each |
|
15908300 |
Nabertherm
HT0627QNA |
64 L |
42730.97€ Each |
|
15928300 |
Nabertherm
HT1227QNA |
128 L |
60739.20€ Each |
|
15948300 |
Nabertherm
HT1627QNA |
160 L |
54617.14€ Each |
|
15968300 |
Nabertherm
HT2727QNA |
276 L |
84128.91€ Each |
|
15988300 |
Nabertherm
HT4527QNA |
450 L |
109555.20€ Each |
|
Description
- Maximum Temperature: 1750 °C
- Dual shell housing with fan cooling for low shell temperatures
- Heating from both sides via molybdenum disilicide heating elements
- High-quality fiber insulation backed by special insulation
- Side insulation constructed with tongue and groove blocks provides for low heat loss to the outside
- Long-life roof insulation with special suspension
- Chain-guided parallel swivel door for defined opening and closing of the door
- Two-door design (front/back) for high-temperature furnaces > HT 276/..
- Labyrinth sealing ensures the least possible temperature loss in the door area
- Reinforced floor as protection for fiber insulation as standard from models HT 16/16 upwards
- Exhaust air opening in the furnace roof
- Heating elements switched via thyristors
- Over-temperature limiter with adjustable cutout temperature for thermal protection class 2 in accordance with EN 60519-2 as temperature limiter to protect the furnace and load
- Defined application within the constraints of the operating instructions
- NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive