missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap Capto Butyl ImpRes

Product Code. 15509271 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 X 5 mL
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15509271 5 X 5 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15509271 Supplier Cytiva Supplier No. 17371912

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HiTrap Capto Butyl ImpRes is a ready-to-use column prepacked with BioProcess Capto Butyl ImpRes resins for screening and small-scale protein purification using hydrophobic interaction chromatography.

  • Prepacked columns for screening of selectivity, binding and elution conditions, as well as small scale purifications.
  • High-throughput chromatography resin for intermediate and polishing steps in a downstream protein purification process.
  • Excellent chemical stability allows cleaning with all standard CIP solutions, including 1 M sodium hydroxide.
  • High binding capacity, high flow rate, and low back pressure reduces process cycle times and improves productivity at process scale.
TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 5 mL
Diameter (Metric) Inner 16 mm
Quantity 5 X 5 mL
Height (Metric) 25 mm
Max. Pressure 5 bar/0.5 MPa/70 psi
Content And Storage 20% ethanol at 4 to 30 °C
Column Type Affinity Column
For Use With (Application) Hydrophobic Interaction Chromatography
compliance-icons
Product Identifier
  • HiTrap Capto Butyl ImpRes 5 ml, 5 x 5 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.