Learn More
- Home
- Products
- Gel Electrophoresis Equipment and Supplies
- Gel Electrophoresis Equipment
- Electrophoresis System Accessories and Replacement Parts
- Hoefer™ Combs for SE 600 Series or SE 400 Series Vertical Electrophoresis Units
Hoefer™ Combs for SE 600 Series or SE 400 Series Vertical Electrophoresis Units
Product Code. 12014707
Change view
Click to view available options
Type:
Adjustable Comb Back
Comb
Preparative Comb
Standard Comb
No. of Wells:
1 Prep, 1 Reference
1 Prep, 2 Reference
10
12
15
20
28
N/A
Thickness (Metric):
0.07 cm
0.1 cm
0.15 cm
N/A
Volume (Metric) Well:
11.5μL/ mm
112μL/ mm
12.4μL/ mm
120μL/ mm
171μL/ mm
183μL/ mm
2.1μL/ mm
2.7μL/ mm
3.1μL/ mm
4.1μL/ mm
4.3μL/ mm
5.7μL/ mm
5.8μL/ mm
6.2μL/ mm
7.7μL/ mm
8.3μL/ mm
8.6μL/ mm
85μL/ mm
90μL/ mm
N/A
Unit Size:
Each
25 product options available for selection
| Product Code. | Type | No. of Wells | Thickness (Metric) | Volume (Metric) Well | unitSize |
|---|---|---|---|---|---|
| 12014707 | Comb | 1 Prep, 2 Reference | 0.07 cm | 85μL/ mm | Each |
| 11579713 | Comb | 12 | 0.1 cm | 7.7μL/ mm | Each |
| 12084867 | Comb | 1 Prep, 2 Reference | 0.15 cm | 171μL/ mm | Each |
| 12074807 | Preparative Comb | 1 Prep, 2 Reference | 0.1 cm | 112μL/ mm | Each |
| 12004877 | Standard Comb | 20 | 0.07 cm | 3.1μL/ mm | Each |
| 10192982 | Standard Comb | 15 | 0.15 cm | 8.6μL/ mm | Each |
| 12069807 | Standard Comb | 20 | 0.15 cm | 6.2μL/ mm | Each |
| 10660036 | Standard Comb | 20 | 0.15 cm | 6.2μL/ mm | Each |
| 12054647 | Standard Comb | 15 | 0.07 cm | 4.3μL/ mm | Each |
| 12074717 | Standard Comb | 10 | 0.07 cm | 6.2μL/ mm | Each |
| 11955348 | Standard Comb | 12 | 0.15 cm | 11.5μL/ mm | Each |
| 11945348 | Standard Comb | 12 | 0.07 cm | 5.8μL/ mm | Each |
| 12064757 | Standard Comb | 10 | 0.15 cm | 12.4μL/ mm | Each |
| 10341135 | Standard Comb | 10 | 0.15 cm | 12.4μL/ mm | Each |
| 10783847 | Adjustable Comb Back | N/A | N/A | N/A | Each |
| 12089857 | Adjustable Comb Back | N/A | N/A | N/A | Each |
| 12069817 | Comb | 1 Prep, 1 Reference | 0.07 cm | 90μL/ mm | Each |
| 12014747 | Comb | 1 Prep, 1 Reference | 0.15 cm | 183μL/ mm | Each |
| 11569723 | Comb | 28 | 0.1 cm | 2.7μL/ mm | Each |
| 12014807 | Comb | 1 Prep, 1 Reference | 0.1 cm | 120μL/ mm | Each |
| 12074757 | Comb | 28 | 0.07 cm | 2.1μL/ mm | Each |
| 12079807 | Comb | 28 | 0.15 cm | 4.1μL/ mm | Each |
| 11539723 | Comb | 20 | 0.1 cm | 4.1μL/ mm | Each |
| 12054867 | Comb | 15 | 0.1 cm | 5.7μL/ mm | Each |
| 10142892 | Comb | 10 | 0.1 cm | 8.3μL/ mm | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Lightweight
- Virtually unbreakable
- Come attached to a standard back which produces maximum well depths of 25 mm, except for the 28-well preparative combs which produce well depths of 15 mm
- Adjustable comb back (available separately, below) converts 25 mm deep combs to 10 or 15 mm depth
Spezifikation
Spezifikation
| For Use With (Equipment) | SE 600, SE 400 Series Vertical Electrophoresis System |
| Type | Comb |
| Material | PTFE |
| No. of Wells | 1 Prep, 2 Reference |
| Thickness (English) | 0.02 in. |
| Thickness (Metric) | 0.07 cm |
| Volume (Metric) Well | 85μL/ mm |
| Width (English) Well | 4.4 in. |
| Width (Metric) Well | 11.3 cm |
| Depth (English) | 0.98 in. |
| Depth (Metric) | 2.5 cm |
| Mehr anzeigen Weniger anzeigen |
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?