Learn More
- Home
- Products
- Gel Electrophoresis Equipment and Supplies
- Gel Electrophoresis Equipment
- Electrophoresis System Accessories and Replacement Parts
- Hoefer™ Combs for SE 600 Series or SE 400 Series Vertical Electrophoresis Units
Hoefer™ Combs for SE 600 Series or SE 400 Series Vertical Electrophoresis Units
Product Code. 12014807
Change view
Click to view available options
Type:
Adjustable Comb Back
Comb
Preparative Comb
Standard Comb
No. of Wells:
1 Prep, 1 Reference
1 Prep, 2 Reference
10
12
15
20
28
N/A
Thickness (Metric):
0.07 cm
0.1 cm
0.15 cm
N/A
Volume (Metric) Well:
11.5μL/ mm
112μL/ mm
12.4μL/ mm
120μL/ mm
171μL/ mm
183μL/ mm
2.1μL/ mm
2.7μL/ mm
3.1μL/ mm
4.1μL/ mm
4.3μL/ mm
5.7μL/ mm
5.8μL/ mm
6.2μL/ mm
7.7μL/ mm
8.3μL/ mm
8.6μL/ mm
85μL/ mm
90μL/ mm
N/A
Unit Size:
Each
25 product options available for selection
| Product Code. | Type | No. of Wells | Thickness (Metric) | Volume (Metric) Well | unitSize |
|---|---|---|---|---|---|
| 12014807 | Comb | 1 Prep, 1 Reference | 0.1 cm | 120μL/ mm | Each |
| 11579713 | Comb | 12 | 0.1 cm | 7.7μL/ mm | Each |
| 12084867 | Comb | 1 Prep, 2 Reference | 0.15 cm | 171μL/ mm | Each |
| 12014707 | Comb | 1 Prep, 2 Reference | 0.07 cm | 85μL/ mm | Each |
| 12074807 | Preparative Comb | 1 Prep, 2 Reference | 0.1 cm | 112μL/ mm | Each |
| 12004877 | Standard Comb | 20 | 0.07 cm | 3.1μL/ mm | Each |
| 10192982 | Standard Comb | 15 | 0.15 cm | 8.6μL/ mm | Each |
| 12069807 | Standard Comb | 20 | 0.15 cm | 6.2μL/ mm | Each |
| 10660036 | Standard Comb | 20 | 0.15 cm | 6.2μL/ mm | Each |
| 12054647 | Standard Comb | 15 | 0.07 cm | 4.3μL/ mm | Each |
| 12074717 | Standard Comb | 10 | 0.07 cm | 6.2μL/ mm | Each |
| 11955348 | Standard Comb | 12 | 0.15 cm | 11.5μL/ mm | Each |
| 11945348 | Standard Comb | 12 | 0.07 cm | 5.8μL/ mm | Each |
| 12064757 | Standard Comb | 10 | 0.15 cm | 12.4μL/ mm | Each |
| 10341135 | Standard Comb | 10 | 0.15 cm | 12.4μL/ mm | Each |
| 10783847 | Adjustable Comb Back | N/A | N/A | N/A | Each |
| 12089857 | Adjustable Comb Back | N/A | N/A | N/A | Each |
| 12069817 | Comb | 1 Prep, 1 Reference | 0.07 cm | 90μL/ mm | Each |
| 12014747 | Comb | 1 Prep, 1 Reference | 0.15 cm | 183μL/ mm | Each |
| 11569723 | Comb | 28 | 0.1 cm | 2.7μL/ mm | Each |
| 12074757 | Comb | 28 | 0.07 cm | 2.1μL/ mm | Each |
| 12079807 | Comb | 28 | 0.15 cm | 4.1μL/ mm | Each |
| 11539723 | Comb | 20 | 0.1 cm | 4.1μL/ mm | Each |
| 12054867 | Comb | 15 | 0.1 cm | 5.7μL/ mm | Each |
| 10142892 | Comb | 10 | 0.1 cm | 8.3μL/ mm | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Lightweight
- Virtually unbreakable
- Come attached to a standard back which produces maximum well depths of 25 mm, except for the 28-well preparative combs which produce well depths of 15 mm
- Adjustable comb back (available separately, below) converts 25 mm deep combs to 10 or 15 mm depth
Specifications
Specifications
| For Use With (Equipment) | SE 600, SE 400 Series Vertical Electrophoresis System |
| Type | Comb |
| Material | PTFE |
| No. of Wells | 1 Prep, 1 Reference |
| Thickness (English) | 0.03 in. |
| Thickness (Metric) | 0.1 cm |
| Volume (Metric) Well | 120μL/ mm |
| Width (English) Well | 4.7 in. |
| Width (Metric) Well | 12.1 cm |
| Depth (English) | 0.98 in. |
| Depth (Metric) | 2.5 cm |
| Show More Show Less |