Description
SE245 Dual-Gel Caster
- For use with SE250, SE260, or MiniVE Vertical Electrophoresis Systems
- Suitable for single concentration and gradient gels
- SE235 4-Gel Caster is for use with SE260 or MiniVE Electrophoresis Systems
- SE 275 4-Gel Caster and SE215 10-Gel Caster are for use with SE250 Electrophoresis System
Specifications
Specifications
| For Use With (Equipment) | SE 250, SE 260 Mini Vertical Gel Electrophoresis Unit |
| Type | Gel Caster |
| Includes | Casting cradle, clamp assemblies, sealing gasket set, cams |
| No. of Gels | 1 to 2 |