Description
SE245 Dual-Gel Caster
- For use with SE250, SE260, or MiniVE Vertical Electrophoresis Systems
- Suitable for single concentration and gradient gels
- SE235 4-Gel Caster is for use with SE260 or MiniVE Electrophoresis Systems
- SE 275 4-Gel Caster and SE215 10-Gel Caster are for use with SE250 Electrophoresis System
Specifications
Specifications
| For Use With (Equipment) | SE 250 Electrophoresis System |
| Type | Gel Caster |
| Includes | Caster body with gasket, face plate with inlet port, 10 x 8cm rectangular Glass plates, 10 x 8cm notched Alumina plates, spring clamps, Wax Paper, space-saver plate, Polycarbonate filler sheets, black Vinyl cap, filler plug set |
| No. of Gels | 4 to 10 |