Learn More
Hoefer™ Hoefer™ Multiple Gel Caster
Enables several gradient or homogenous gels to be cast simultaneously. Cast homogenous or gradient gels.
Brand:  Hoefer™ SE615
Additional Details : Weight : 0.80000kg
Product Code. 12064707
  • 919.00€ / Each
Estimated Shipment
26-02-2021
Add to basket
Description
Description
  • Casts up to ten 16cm long gels
  • Easy to use and leak free
  • Cast homogenous or gradient gels
    • Pour homogenous gels directly into the top of the casting chamber
    • Pump gradient gels through an inlet port at the bottom
Includes:
  • Caster Body with Gasket
  • Spacer-Mate–Alignment Template
  • Face Plate
  • Thumbscrews–2 pcs
  • Clamps–4 pcs
  • Filler Plug Set
  • Glass Plates
  • Black Vinyl Cap 
  • Filter Sheets–5 pcs
  • Gel Seal 
  • Wax Paper–100 Sheets, Precut
  • Bubble Level
Specifications
Specifications
Ten Gel Caster
Multiple Gel Caster
Includes 20 Glass Plates, 18 x 16cm