missing translation for 'onlineSavingsMsg'
Learn More

Sperian™ Honeywell™ Viking Noise Blocking Ear Muffs

Product Code. p-8010186
Change view
Click to view available options
Color:
Black, Red
Black, Silver
Unit Size:
Each
Pack of 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
12685033 Black, Silver Each
12635583 Black, Red Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12685033 Supplier Sperian™ Supplier No. 1011170

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Honeywell™ Viking Noise Blocking Ear Muffs feature a multi-position headband for positioning over the head, behind the head, or under the chin. Attached elastic headband strap allows better positioning behind the head or under the chin. Suitable for electrical environments.

  • Alternative to cap-mounted earmuffs – wear with hard hats, face shields, respirators and other PPE
  • Patented Air Flow Control™ technology delivers optimal attenuation across all frequencies without increasing earcup size or weight
  • Inner-ventilated headband minimizes pressure on the head, breathes easier in warm/humid climates [V2 and V3 only]
  • Non-deforming outer headband withstands rough treatment in the toughest workplaces
  • Snap-in ear cushions make replacement quick and easy
TRUSTED_SUSTAINABILITY

Specifications

Color Black, Silver
Certifications/Compliance PPE CAT II, ISO 9001/2000, 29 CRF 1910.95, 2003/10/EC, 89/686/EEC
For Use With (Application) Noise Blocking Ear Muff
Product Type Hearing Protection
Dielectric Yes
Style Multi-position
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.