Learn More
UVP Lamp UVM-24 EL series UV supplied with 2 x 302nm tubes 230V 50Hz 4W UVP
Brand:  UVP 95026802
Additional Details : Weight : 1.50000kg
Product Code. 12862969
Quantity Price
1 358.00€ / Each
6 308.24€ / Each
Estimated Shipment
17-02-2021
Add to basket