Learn More
BRAND™ LDPE Pasteur Pipettes
Product Code. p-8000113 Shop All Brand Products
Change view
Click to view available options
Length (Metric):
144 mm
150 mm
152 mm
Volume (Metric):
1 mL
2 mL
3 mL
3.5 mL
4 mL
Diameter (Metric) Outer:
1 mm
2.8 mm
3 mm
3.2 mm
3.3 mm
3.4 mm
Drops/mL:
21 to 28
22 to 26
25 to 27
25 to 30
50 mL
60 to 75
Unit Size:
Pack of 500
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Volume (Metric) Diameter (Metric) Outer Drops/mL unitSize
15443594 144 mm 1 mL 1 mm 50 mL Pack of 500
10646341 152 mm 2 mL 3.3 mm 22 to 26 Pack of 500
10562291 152 mm 3.5 mL 3.2 mm 21 to 28 Pack of 500
10222661 150 mm 1 mL 3.4 mm 25 to 30 Pack of 500
10100531 152 mm 3 mL 2.8 mm 25 to 27 Pack of 500
10721963 150 mm 4 mL 3 mm 60 to 75 Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15443594 Supplier BRAND™ Supplier No. 747775
Only null left
Add to basket
Edge
Add to basket
LDPE Pasteur Pipettes are ideal for aliquots with very good reproducibility of the number of drops per milliliter.
  • Disposable
  • Very good reproducibility of the number of drops per mL
  • Ideal for aliquots
  • Can be deep-frozen in filled state or changed into a closed vessel by heat-sealing the tip
  • Integrated pipetting bulb depresses easily, minimize fatigue from frequent pipetting
  • PE-LD material
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Outer 1 mm
Material LDPE
Drops/mL 50 mL
Quantity 500 Pack
Top Style Bulb Shaped
Volume (Metric) 1 mL
Length (Metric) 144 mm
Type Pasteur Pipet