Learn More
Brannan™ LO-tox™ Lab Thermometer
LO-tox™ lab thermometer without calibration certificate.
3.35€ - 61.50€
Product Code | Brand | Temperature Range (Metric) | Subdivision | Immersion Type | Length (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Temperature Range (Metric) | Subdivision | Immersion Type | Length (Metric) | Price | Quantity & Availability | ||
13008593
|
Brannan™
44/800/8 |
-10°C to +50°C | 0.5°C | Partial Immersion | 305 mm |
13.30€
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
13098583
|
Brannan™
44/801/8 |
-35°C to +50°C | 1°C | Partial Immersion | 305 mm |
4.45€
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
13018593
|
Brannan™
44/802/8 |
-20°C to +110°C | 1°C | Partial Immersion | 305 mm |
11.20€
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||
13028593
|
Brannan™
44/803/8 |
-20°C to +150°C | 1°C | Partial Immersion | 305 mm |
3.95€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
11907341
|
Brannan™
44/804/8 |
-10°C to +260°C | 1°C | Partial Immersion | 405 mm |
4.60€
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
13038583
|
Brannan™
44/805/8 |
-10°C to +210°C | 1°C | Partial Immersion | 405 mm |
21.45€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
12309359
|
Brannan™
44/808/8 |
-10°C to +50°C | 0.5°C | Total Immersion | 305 mm |
12.00€
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||
12399349
|
Brannan™
44/809/8 |
-35°C to +50°C | 1°C | Total Immersion | 305 mm |
4.45€
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||
13078583
|
Brannan™
44/810/8 |
-20°C to +110°C | 1°C | Total Immersion | 305 mm |
12.50€
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
13088583
|
Brannan™
44/811/8 |
-20°C to +150°C | 1°C | Total Immersion | 305 mm |
13.30€
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
12389349
|
Brannan™
44/812/8 |
-10°C to +210°C | 1°C | Total Immersion | 405 mm |
4.70€
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
13068593
|
Brannan™
44/814/0 |
-10°C to +330°C | 2°C | Partial Immersion | 305 mm |
19.60€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
12319359
|
Brannan™
44/815/0 |
-10°C to +300°C | 1°C | Partial Immersion | 305 mm |
19.60€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
13038603
|
Brannan™
44/820/0 |
-1°C to +51°C | 0.1°C | Partial Immersion | 405 mm |
16.10€
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
13008603
|
Brannan™
44/821/0 |
-1°C to +51°C | 0.1°C | Total Immersion | 405 mm |
16.10€
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||
13048603
|
Brannan™
44/822/0 |
-1°C to +51°C | 0.2°C | Partial Immersion | 405 mm |
15.60€
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
13018603
|
Brannan™
44/823/0 |
-1°C to +51°C | 0.2°C | Total Immersion | 405 mm |
15.60€
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
13058603
|
Brannan™
44/824/0 |
-10°C to +101°C | 0.2°C | Partial Immersion | 405 mm |
16.55€
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
12339359
|
Brannan™
44/825/0 |
-1°C to +101°C | 0.2°C | Total Immersion | 405 mm |
16.55€
Each |
Estimated Shipment: 21-06-2024
Log in to see stock. |
||
13038593
|
Brannan™
44/830/7 |
-10°C to +110°C | 1°C | Total Immersion | 155 mm |
9.35€
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||
13048593
|
Brannan™
44/850/7 |
-10°C to +50°C | 1°C | Total Immersion | 115 mm |
9.80€
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
15482695
|
Brannan™
44/819/0 |
-10°C to +360°C | 2°C | Partial Immersion | 305 mm |
61.50€
Pack of 10 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
15442695
|
Brannan™
44/810/9 |
-20°C to +110°C | 0.5°C | Total Immersion | 305 mm |
33.35€
Pack of 10 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
15432695
|
Brannan™
44/826/8 |
-10°C to +360°C | 2°C | Total Immersion | 305 mm |
59.15€
Pack of 10 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
15689025
|
Brannan™
44/802/9 |
-20°C to +110°C | 0.5°C | Partial Immersion | 305 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||
Description
Read temperatures quickly and accurately with blue spirit against a white background. Brannan™ Lo-Tox™ Spirit-filled Total Immersion Thermometer is manufactured to traceable standards and includes a blue non-roll top and safety suspension ring.
- General purpose/stirring thermometer with a low toxic, environmentally friendly filling
- Easy to read against background
- Each thermometer is manufactured to traceable standards
Specifications
Glass |