missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Interscience Scan 100 Manual Colony Counter
Description
Manual colony counter with LEDs Lighting, USB data transfer, black/white background (100-240V) delivered with:
- 1 white background plate
- 1 dia. 90 mm Petri dish adapter
- 1 black marker pen
- 1 power cable
- 1 USB cable
- 1 quick user guide
Specifications
Specifications
| Product Type | Manual colony counter |
| Display Type | LCD |
| Description | Scan 100 enables the accurate counting of colonies on any type of media. The integrated USB key enables to export the results to guarantee traceability. Diameter of Petri Dishes : dia. 55, 65, 90 and 150 mm |
| Color | Gray (STAINLESS STEEL) |
| Detection Method | Manual colony counter (from 0 to 1998 CFU) |
| Field | For all culture supports: Petri dishes: dia. 55 - 150 mm: PetriFilmTM, Compact DryTM, MC-Media PadsTM, easyPlateTM, filtration membranes |
| Magnification | The magnifying lens is in option (Ref. 435 001) |
| Material | Stainless Steel, Neoprene cell |
| Memory | Data export to LIMS, via USB, as CSV spreadsheet (ExcelTM) or text (WordTM) format |
| Model | Scan 100 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?