Learn More
Thermo Scientific™ MaxQ™ 4000 Gassing Manifold
Enable direct gassing into flasks for anaerobic cultures with the Thermo Scientific™ MaxQ™ 4000 Gassing Manifold.
Brand: Thermo Scientific™ SHK4000-8GM
Additional Details : Weight : 10.00000kg
Specifications
| Gassing manifold with 8 outlets | |
| MaxQ 4000 | |
| MaxQ 4000 |
| Gassing manifold with 8 outlets | |
| Gassing Manifold |