missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ MES, 1.0M buffer soln., pH 5.5

67.00€ - 104.00€

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.248
MDL Number MFCD00283848
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
PubChem CID 16218417
IUPAC Name 2-morpholin-4-ylethanesulfonic acid;hydrate
SMILES C1COCCN1CCS(=O)(=O)O.O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15445599 SDS View Documents Alfa Aesar™
J63341.AK
250mL 67.00€
250mL View alternative products
Save Up to 
»
EU Stock 3
Add to basket
 
15455599 SDS View Documents Alfa Aesar™
J63341.AP
500mL 104.00€
500mL View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 13-07-2020
Add to basket
 
Description

Description

Applications
MES is used as a buffering agent in biology and biochemistry. It is utilized for resolving very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

145224-94-8
213.248
MIIIXQJBDGSIKL-UHFFFAOYSA-N
2-morpholin-4-ylethanesulfonic acid;hydrate
C6H15NO5S
MFCD00283848
16218417
C1COCCN1CCS(=O)(=O)O.O

Specifications

MES buffer soln.
C6H15NO5S
1.0M buffer soln.
MIIIXQJBDGSIKL-UHFFFAOYSA-N
2-morpholin-4-ylethanesulfonic acid;hydrate
16218417
5.5
145224-94-8
MFCD00283848
Miscible with water.
C1COCCN1CCS(=O)(=O)O.O
213.248
Liquid
250mL