missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ MES, 1.0M buffer soln., pH 8.5

59.10€ - 91.60€

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.248
MDL Number MFCD00283848
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
PubChem CID 16218417
IUPAC Name 2-morpholin-4-ylethanesulfonic acid;hydrate
SMILES C1COCCN1CCS(=O)(=O)O.O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15405039 SDS View Documents Alfa Aesar™
J62720.AK
250mL 59.10€
250mL View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 13-07-2020
Add to basket
 
15415039 SDS View Documents Alfa Aesar™
J62720.AP
500mL 91.60€
500mL View alternative products
Save Up to 
»
EU Stock 1
Add to basket
 
Specifications

Chemical Identifiers

145224-94-8
213.248
MIIIXQJBDGSIKL-UHFFFAOYSA-N
2-morpholin-4-ylethanesulfonic acid;hydrate
C6H15NO5S
MFCD00283848
16218417
C1COCCN1CCS(=O)(=O)O.O

Specifications

MES buffer soln.
C6H15NO5S
1.0M buffer soln.
C1COCCN1CCS(=O)(=O)O.O
213.248
Liquid
250mL
145224-94-8
MFCD00283848
MIIIXQJBDGSIKL-UHFFFAOYSA-N
2-morpholin-4-ylethanesulfonic acid;hydrate
16218417
8.5