Learn More
Esculape Laboratoire™ Mix Eyestation
Description
- Mixed preservative with tap water
- ontinuous flushing with 0.4 gallons per minute for 15 minutes
- Hands-free operation allows user to hold both eyes open simultaneously
Specifications
Specifications
| For Use With (Application) | Washing the Eyes in the Event of Splashes of Acids or Bases |
| Includes | Eye wash bottle with 200 mL neutral pH solution; Eye wash bottle with 500 mL of ophthalmic solution; Box for wall mounting with mirror and user manual |
| Rinse Time | 7 min. |