missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves, 13X, powder

Product Code. p-7080729
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424203 250 g 250g
11414203 1000 g 1000g
11434203 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424203 Supplier Thermo Scientific Alfa Aesar Supplier No. A10378.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 63231-69-6

They are used as catalyze carrier, co adsorption of CO2 and H2O and H2O, H2O and H2S, as desiccant for medical and air compressor systems. They are normally used in gas phase and liquid phase applications. It finds its a major applications in commercial gas drying, air plant feed purification (simultaneous H2O and CO2 removal) and liquid hydrocarbon/natural gas sweetening (H2S and mercaptan removal).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63231-69-6
MDL Number MFCD00131613
IUPAC Name Molecular sieves 13X

Specifications

Sensitivity Hygroscopic
Quantity 250 g
Physical Form Powder
Solubility Information Insoluble in water.
Chemical Name or Material Molecular sieves 13X

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.