missing translation for 'onlineSavingsMsg'
Learn More

Nabertherm Muffle Furnace, 1100 L Series with C450 Controller

For daily laboratory use due to excellent workmanship, advanced design, and high level of reliability.

3013.20€ - 5724.00€


Products 3
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15426672 View Documents Nabertherm
L-031K1TN
3 L 3013.20€
Each View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 12-09-2019
Add to basket
 
15416702 View Documents Nabertherm
L-091K1TN
9 L 3855.60€
Each View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 12-09-2019
Add to basket
 
15446722 View Documents Nabertherm
L-241K1TN
24 L 5724.00€
Each View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 03-10-2019
Add to basket
 
Description

Description

  • Maximum Temperature: 1100 °C
  • Heating from two sides by ceramic heating plates (heating from three sides for muffle furnace L 24/11)
  • Ceramic heating plates with integral heating element which is safeguarded against fumes and splashing, and easy to replace
  • Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
  • Housing made of sheets of textured stainless steel
  • Dual shell housing for low external temperatures and high stability
  • Flap door can be used as work platform or lift door
  • Adjustable air inlet integrated in door (see illustration)
  • Exhaust air outlet in rear wall of furnace
  • Solid state relays provide for low-noise operation
  • Defined application within the constraints of the operating instructions
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive