Learn More
Nabertherm Muffle Furnace L 1200 Series
For daily laboratory use due to excellent workmanship, advanced design, and high level of reliability.
3574.80€ - 4957.20€
Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15456672 View Documents Nabertherm
L-031K2TN
3 L 3574.80€
Each View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 12-09-2019
Add to basket
15446712 View Documents Nabertherm
L-151K2TN
15 L 4957.20€
Each View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 12-09-2019
Add to basket
Description
Description
  • Maximum Temperature: 1200 °C
  • Heating from two sides by ceramic heating plates
  • Ceramic heating plates with integral heating element which is safeguarded against fumes and splashing, and easy to replace
  • Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
  • Housing made of sheets of textured stainless steel
  • Dual shell housing for low external temperatures and high stability
  • Flap door can be used as work platform or lift door
  • Adjustable air inlet integrated in door (see illustration)
  • Exhaust air outlet in rear wall of furnace
  • Solid state relays provide for low-noise operation
  • Defined application within the constraints of the operating instructions
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive