Learn More
Nabertherm Muffle Furnace LT 1200 Series with B410 Controller
For daily laboratory use due to excellent workmanship, advanced design, and high level of reliability.
4125.60€ - 4330.80€
| Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
| 15496692 |
Nabertherm
L-091H2RN |
9 L |
4125.60€ Each |
from Supplier 03-10-2019 |
15406712 |
Nabertherm
L-151H2RN |
15 L |
4330.80€ Each |
from Supplier 03-10-2019 |
Description
- Maximum Temperature: 1200 °C
- Heating from two sides by ceramic heating plates
- Ceramic heating plates with integral heating element which is safeguarded against fumes and splashing, and easy to replace
- Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
- Housing made of sheets of textured stainless steel
- Dual shell housing for low external temperatures and high stability
- Lift door with hot surface facing away from the operator
- Adjustable air inlet integrated in door (see illustration)
- Exhaust air outlet in rear wall of furnace
- Solid state relays provide for low-noise operation
- Defined application within the constraints of the operating instructions
- NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive