Learn More
Nabertherm Muffle Furnace LT 1200 Series with B410 Controller
For daily laboratory use due to excellent workmanship, advanced design, and high level of reliability.
4125.60€ - 4330.80€
Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15496692 View Documents Nabertherm
L-091H2RN
9 L 4125.60€
Each View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 03-10-2019
Add to basket
15406712 View Documents Nabertherm
L-151H2RN
15 L 4330.80€
Each View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 03-10-2019
Add to basket
Description
Description
  • Maximum Temperature: 1200 °C
  • Heating from two sides by ceramic heating plates 
  • Ceramic heating plates with integral heating element which is safeguarded against fumes and splashing, and easy to replace
  • Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
  • Housing made of sheets of textured stainless steel
  • Dual shell housing for low external temperatures and high stability
  • Lift door with hot surface facing away from the operator
  • Adjustable air inlet integrated in door (see illustration)
  • Exhaust air outlet in rear wall of furnace
  • Solid state relays provide for low-noise operation
  • Defined application within the constraints of the operating instructions
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive