Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Square PETG Platinum Certified Clean Media Bottle

Click to view available options
Capacity (Metric):
5 mL
20 mL

Thermo Scientific™ Nalgene™ PETG Diagnostic Bottles safe for food and beverage use

Products are washed with USP Water for Injection and handled post-washing in Class 5 clean rooms with Class 4 packaging/inspection hoods to produce these very low particulate products. Certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready-to-use products with a Sterility Assurance Level of 10-6 inside the container/closure, including two heat-sealed sterile barriers to protect product prior to use

  • Sterile to 10-6 SAL
  • Non-pyrogenic and non-cytotoxic
  • Leakproof High Density Polyethylene (HDPE) closures.
  • Serum Vials are packed in shrink-wrapped trays with closures attached.
  • Manufactured in compliance with ISO 13485:2003 quality requirements
Certifications/Compliance ISO 13485:2003, 21CFR177.1520, USP Class VI
Capacity (Metric) 5 mL
Capacity (English) 0.16 oz.
Color Clear
Diameter (Metric) Inner Neck 11 mm
For Use With (Application) Sterile Sampling, Storage, and Shipment
Material PETG
No. per Case 416
No. per Pack 52
Weight (English) 9 lb.
Width (English) 12.5 in.
Autoclavable No
Closure Size 20 mm
Diameter (Metric) 22 mm
Height (Metric) 46 mm
Show More Show Less