Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences DURAN™ Original GL 45 Lab Bottles - Complete with High Temperature Closures
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1 L
2 L
5 L
10 L
Height (Metric):
143mm
415mm
105mm
181mm
230mm
265mm
335mm
Description
DURAN™ 3.3 borosilicate bottles are in comformanceUSP/EP Type 1 providing purity and exceptional performance. DURAN™ borosilicate 3.3 glass, renowned for its excellent thermal performance and high resistance to chemical attack. High Temperature Cap is ideal for applications to 180°C. Permanent Retrace Code on each bottle for full manufacturing lot traceability.
Specifications
| Capacity (Metric) | 100 mL |
| Color | Clear |
| Graduated | Yes |
| For Use With (Application) | High temperature storage, filling, transfer of high value or aggressive solutions. Autoclaving liquid media. Storage of corrosive reagents. Sampling. |
| Includes | ETFE Pour Ring |
| Material | Borosilicate Glass 3.3 |
| Autoclavable | Yes |
| Closure Size | GL 45 |
| Graduations | Permanent graduations |
| Height (Metric) | 105mm |