Learn More
Parker domnick hunter™ Clear Desiccant Cartridge for Hydrogen Gas Generators
Contains a combination of white desiccant beads and orange indicating beads that change to clear as they become saturated
Brand:  Parker domnick hunter™ 604970050
Additional Details : Weight : 0.80000kg
Product Code. 12517005
  • 984.50€ / Each
Estimated Shipment
10-10-2019
Add to basket
Description
Description
  • Contains a combination of white desiccant beads and orange indicating beads that change to clear as they become saturated
  • Disposable cartridge cannot be heat regenerated
  • Vacuum sealed in aluminum foil
  • Fits all domnick hunter™ UHP Hydrogen Generator models
Specifications
Specifications
Hydrogen gas generators uhp-20h, uhp-40h and uhp-60h
Hydrogen gas generators uhp-20h, uhp-40h and uhp-60h
Combination of white desiccant beads and orange indicating beads that change to clear as they become saturated
Desiccant Cartridge