Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Pierce™ 0.1% Formic Acid (v/v) in Water, LC-MS Grade
Click to view available options
Quantity:
1 L
4 x 1 L
Description
Thermo Scientific™ Pierce 0.1% Formic Acid in Water is an LC/MS-grade preparation with high purity and low UV-absorptivity that is ideal for HPLC and mass spectrometry applications.
Additional Information and Online Ordering »
Highlights:
Validated
– tested more than 20 ways to ensure maximal LC/MS sensitivity
Purified
– processed, filtered and sealed to extend LC/MS column life
Convenient
– packaged to eliminate variability and reduce handling
Specifications
| Reagent Type | Solvent Mixture |
| Form | Liquid |
| Product Line | Pierce™ |
| Content And Storage | Store in a cool, dry, and well-ventilated area. Protect from direct sunlight. |
| Quantity | 4 x 1 L |