Learn More
Thermo Scientific™ Pierce™ Protein-Free (TBS) Blocking Buffer
Block membranes, plates and other samples with these ready-to-use PBS and TBS formulations of a non-protein compound to yield extremely low background.
44.58€ - 201.00€
Products 2
Product Code Brand Description Quantity Price Quantity & Availability  
Product Code Brand Description Quantity Price Quantity & Availability  
10269613 View Documents Thermo Scientific™
37570
Protein-Free (TBS) Blocking Buffer 1 L 201.00€
1 L View alternative products
Save Up to 
»
EU Stock 43
Add to basket
13484299 View Documents Thermo Scientific™
37585
Protein-Free (TBS) Blocking Buffer 100 mL 44.58€
100mL View alternative products
Save Up to 
»
EU Stock 7
Add to basket
Description
Description
Traditional blocking buffers contain proteins that can cross-react with immunodetection systems, resulting in high background and reduced signal. Typical commercial alternatives to protein-based blocking agents are ineffective. Pierce Protein-Free Blocking Buffers are devoid of protein while remaining highly effective at blocking plates, membranes and other surfaces for ELISA, Western blotting, glass slide arrays and other applications.
Highlights:
  • Protein-free blocker – minimizes or eliminates cross-reactivity associated with protein-based blocking buffers
  • Application-compatible – effective in all kinds of protein detection systems, including Western blots (membranes), ELISA (microplates) and arrays (coated glass slides)
  • Streptavidin-friendly – absolutely free of biotin; no possible interference with avidin-biotin detection systems
  • High-performance – optimized and validated in many protein methods to provide high signal-to-noise ratio (i.e., no quenching of specific binding and signal but eliminating nonspecific binding and background)
  • Convenient – supplied as ready-to-use 1X formulations in TBS and PBS with and without 0.05% Tween™-20