Learn More
Thermo Scientific™ Pierce™ Water, LC-MS Grade
Description
Thermo Scientific™ Pierce Water, LC/MS Grade is ultapure water for formulation of solvents for HPLC
Thermo Scientific™ Pierce Water, LC/MS Grade is ultapure water for formulation of solvents for HPLC.
Additional Information and Online Ordering »
An ultrapure, LC/MS-grade preparation with low UV-absorptivity that makes it suitable and reliable for use in HPLC and mass spectrometry applications.
Thermo Scientific Pierce Water, LC/MS Grade is specially purified by a proprietary method and treated to ensure lot-to-lot consistency with a low UV absorbance to provide you with most sensitive detection across all wavelengths. Pierce Water is 0.2-micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen atmoshere with Teflon TFE-lined fluorocarbon caps for ultimate protection.
Highlights:
- Purity and quality validated with 30 tests for use in LC/MS applications
- 0.2-micron filtered, packed in solvent-rinsed amber glass bottles
- Sealed under nitrogen atmosphere to eliminate absorption of unknown atmospheric gases
Product Details:
General properties of water
- Molecular formula: H2O
- Molecular Weight: 18.01
- CAS number: 7732-18-5
Pierce Water, LC/MS Grade is specifically purified and tested to the highest specifications to ensure the integrity of your data, maximize sensitivity in your assay and prolong the life of your equipment.
Specifications
| Physical Form | Liquid |
| Reagent Type | Solvent (Pure) |
| Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area. |
| Product Line | Pierce™ |
| Quantity | 1L |