Learn More
Bel-Art™ SP Scienceware™ Wilmad™ Pipet Bulb
Description
- Latex rubber
- 6mm dia. bore
Specifications
Specifications
| Length (Metric) | 34.93 cm |
| Material | Latex Rubber |
| Description | Latex bulb |
| Diameter (Metric) Inner | 6 mm |
| Quantity | 50 Pack |
| Model | 804 |