missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Polypropylene Conical 384 Deepwell Protein LoBind Plate

LoBind, free from DNA, DNase, RNase and PCR inhibitors

743.00€ - 2004.00€

Specifications

No. of Wells 384
Certified SBS, IVD, EN ISO 9001/2/3 and EN 13485
Dnase Free Yes
RNase Free Yes
Volume (Metric) 200 μL
View More Specs

Products 2
Product Code Brand No. per Pack Price Quantity & Availability  
Product Code Brand No. per Pack Price Quantity & Availability  
10748483
View Documents
Eppendorf™
0030524101
40
743.00€
Pack of 40
View alternative products
Save Up to 
»
Estimated Shipment
19-01-2021
Add to basket
 
10583984
View Documents
Eppendorf™
0030528107
120
2004.00€
Pack of 120
View alternative products
Save Up to 
»
Estimated Shipment
20-01-2021
Add to basket
 
Description

Description

The Eppendorf polypropylene deep well plates combine a new well design with a sophisticated color coding system with ease of reading and orientation. The well geometry allows high speed pipetting without splashing, excellent mixing and high speed centrifugation, which accelerates your manual and automated work processes. The new polypropylene plates fulfill all current SBS, IVD, EN ISO 9001/2/3 and EN 13485 guidelines.
  • Recovermax - concaved round bottom well shape with smoothed edges
  • Optitrack - color framed plates with alphanumeric labelling
  • High chemical resistance
  • G-safe - highest centrifugation stability
  • LoBind - free of surface coating e.g., silicone
  • Free of DNA, DNase, RNase and PCR* inhibitor (PCR* clean)
  • LoBind Protein - minimum protein loss (less than 3% BSA 1μg/mL)
  • LoBind DNA - less than 1% loss of DNA/RNA at low concentrations and critical conditions (high salt, short DNA fragments)
  • Autoclavable
  • Proofed automatic usability
  • Conform to SBS standard
  • Optionally available with barcode
Specifications

Specifications

384
Yes
200 μL
384-Well Protein LoBind Deepwell Plates
SBS, IVD, EN ISO 9001/2/3 and EN 13485
Yes
V
SDS