missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Polypropylene Rods for Draining Racks

Product Code. p-8002468
Change view
Click to view available options
Product Type:
Rod
Rod Extension
Size:
Dia. x L: 10 x 100 mm
Dia. x L: 12 x 150 mm
Dia. x L: 6 x 60 mm
Color:
Blue
Transparent
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Size Color unitSize
10579801 Rod Dia. x L: 6 x 60 mm Blue Each
10007560 Rod Dia. x L: 10 x 100 mm Blue Each
10352891 Rod Extension Dia. x L: 12 x 150 mm Transparent Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10579801 Supplier Buerkle™ Supplier No. 96400421
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Buerkle™ Polypropylene Rods for Draining Racks can be replaced and have their positions changed.

  • The rods can be replaced and have their positions can be changed
  • The extension can only be used with the rods of 100 mm length
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Material Polypropylene
For Use With (Equipment) Draining Rack
No. per Pack 1
Size Dia. x L: 6 x 60 mm
Length (Metric) 60 mm
Diameter (Metric) Outer 6 mm
Product Type Rod
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.