Learn More
Fisherbrand™ Portable Balance
Portable Balance with a slim, stackable design provides basic weighing functionality and competitive performance.
47.30€ - 58.40€
Specifications
Display Type | LCD |
---|---|
For Use With (Application) | Weighing |
Includes | 3 AA (LR6) Batteries |
Stabilization Time | 2.000 sec. |
Temperature Range (English) | 64°F to 77°F |
Product Code | Brand | Capacity | Model | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Model | Price | Quantity & Availability | ||||
15939345
|
Fisherbrand™
|
220 g | FCS221 |
47.30€
Each |
In Stock
Log in to see stock. |
||||
15949345
|
Fisherbrand™
|
620 g | FCS621 |
58.40€
Each |
In Stock
Log in to see stock. |
||||
15909355
|
Fisherbrand™
|
2200 g | FCS2200 |
51.00€
Each |
In Stock
Log in to see stock. |
||||
15919355
|
Fisherbrand™
|
5200 g | FCS5200 |
58.40€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
||||
Description
- Intuitive two-button design makes for easy operation
- Slim, space-saving design allows for stackable storage
- Enlarged weighing platform can adequately handle large samples and a variety of objects
Specifications
LCD | |
3 AA (LR6) Batteries | |
64°F to 77°F | |
2 Years | |
5 in. | |
5.2 in. | |
0.9 lb. |
Weighing | |
2.000 sec. | |
18°C to 25°C | |
205 x 140 x 41 mm (8.1 x 5.5 x 1.6 in.) | |
125 mm | |
132 mm | |
0.4 kg |