missing translation for 'onlineSavingsMsg'
Learn More

Esculape Laboratoire™ Eyewash Bottles for First Aid

Product Code. p-8001254
Change view
Click to view available options
Volume (Metric):
200 mL
500 mL
Composition:
Sterile Isotonic Solution
Sterile Ophthalmic Solution
Includes:
200 mL Blue Neutral pH Solution, 50 0mL Bottle Green Sterile Ophthalmic Solution
Cap
Cap, Storage, Carrying Case
Quantity:
10 Pk.
12 Pk.
Ea.
Unit Size:
Each
Pack of 10
Pack of 12
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Composition Includes Quantity unitSize
11769102 200 mL Sterile Isotonic Solution Cap, Storage, Carrying Case Ea. Each
11749522 500 mL Sterile Ophthalmic Solution Cap 12 Pk. Pack of 12
11759522 200 mL Sterile Ophthalmic Solution 200 mL Blue Neutral pH Solution, 50 0mL Bottle Green Sterile Ophthalmic Solution Ea. Each
11891553 200 mL Sterile Ophthalmic Solution Cap 10 Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11769102 Supplier Esculape Laboratoire™ Supplier No. 8205250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use Esculape™ Eyewash Bottles for First Aid has a carrying case with a belt loop for portabilitiy.

  • Storage case
  • Contain sterile isotonic solution
  • Ergonomic

Specifications

Product Type Eyewash Bottle
Composition Sterile Isotonic Solution
For Use With (Application) Portable Eyewash
Includes Cap, Storage, Carrying Case
Quantity Ea.
Sterility Sterile
Volume (Metric) 200 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.