missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Plating pH Portable Meter

Product Code. 11627270
Change view
Click to view available options
Dimensions (L x W x H):
152 x 58 x 30 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
11627270 152 x 58 x 30 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11627270 Supplier Hanna Instruments™ Supplier No. HI99131
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Easy to clean and prevents solids from collecting on the sensor. Hanna Instruments™ Plating pH Portable Meter is a pre-amplified, double junction pH probe with a recessed flat tip. The titanium body functions as a potential matching pin for increased stability and extended sensor life.

The HI99131 is a waterproof, portable pH and temperature meter specifically designed for use in plating baths. Titanium construction provides an unbreakable structure and allows the transfer of heat to the internal temperature sensor for rapid temperature compensation. The maintenance-free, gel-filled electrode does not require fill solution.

  • Automatic Temperature Compensation
  • One or two-point automatic calibration
  • BEPS - Alerts the user of low battery power that could adversely affect readings
  • Compact, heavy-duty, and waterproof
  • Battery % displayed on start-up
  • HELP feature, displays tutorial messages on LCD

Specifications

Includes HI62911D pH probe, HI70004 pH 4.01 buffer solution sachet (20 mL), HI70007 pH 7.01 buffer solution sachet (20 mL), HI700661 electrode cleaning solution sachets (2 x 20 mL), batteries, instructions, and hard carrying case
Accuracy (Temperature) ±0.5°C (up to 60°C); ±1.0°C (outside), ±1°F (up to 140°F);
Accuracy (pH) ±0.02
Calibration Automatic
Range (pH) -2 to 16
Resolution (pH) 0.01
Calibration Points 1 or 2 with 2 sets of standard buffers (pH 4.01, 7.01, 10.01 or pH 4.01, 6.86, 9.18)
Temperature Range (Metric) -5.0°C to +105.0°C
Temperature Range (English) 23.0°F to 221.0°F
Resolution (Temperature) 0.1°C/°F
Temperature Probe Calibration HI62911D titanium body, pre-amplified pH probe with internal temperature sensor, DIN connector and 1 m (3.3’) cable (included)
Display Type LCD
Inputs 5-pin DIN
IP Rating IP67
Dimensions (L x W x H) 152 x 58 x 30 mm
Weight (Metric) 205 g
Weight (English) 7.2 oz.
Warranty 2 Years (6 Months Probe)
Material Titanium Body
Model HI-99131
Power Supply 4 x 1.5V
Probe Type HI62911D titanium body, pre-amplified pH probe with internal temperature sensor, DIN connector and 1m cable
Relative Humidity Range Max 100%
Detection Range -2 to 16 pH
Temperature Compensation Automatic from -5.0 to 105.0°C (23 to 221°F)
Temperature (English) Operating 32°F to 122°F
Temperature (Metric) Operating 0°C to 50°C
Battery Type 3 x 1.5V AAA
Temperature Sensor Yes
Type Portable pH meter
Battery Life 1200 hr.
Length (Metric) 150 mm
Height (Metric) 28 mm
Depth (Metric) 80 mm
Electrical Requirements 4 x 1.5V
Voltage 4 x 1.5 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.