Learn More
Fisherbrand™ Power Supply, Mini 300V Plus
Provides up to 300V (volts) and 400mA of power. Fisherbrand™ Power Supply, 300V Mini Plus includes dual voltage compatibility.
Brand:  Fisherbrand™
Additional Details : Weight : 1.45700kg
Product Code. 12643546
  • 345.00€ / Each
In stock, ready to ship: 8
Add to basket
Description
Description
  • Enhanced built-in safety features
  • Three digit LED display
  • Alarm function
  • Wipe clean polycarbonate housing
Specifications
Specifications
60W
300V
3-digit LED
140 x 191 x 84mm
10 to 300
10 to 400mA
100 to 240V, 50/60Hz