Learn More
Macherey-Nagel™ Protino GST / 4B FPLC 1 mL columns for GST-tag Protein Purification
Product Code. 12939934
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12939934 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12939934 Supplier Macherey-Nagel™ Supplier No. 745510.1
Best choice for cost effective GST-tag protein purification.
  • MPLC, FPLC, peristaltic pump, or syringe
Specifications
Capacity (Metric) 1 mL
Concentration 0.1 M acetate, pH 4, 0.1 M NaOH, 70% ethanol, or 6 M guanidine hydrochloride
Quantity 1 mL
Type Column
Sample Volume 1 mL
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: ethanol