Learn More
Kimberly-Clark™ WypAll™ Critical Clean™ Colour Coded Cleaning Cloths, Red
Product Code. p-8004703
Change view
Click to view available options
Dimensions (L x W):
41.6 x 33 cm (16.4 x 13 in.)
41.8 x 24.7 cm (16.5 x 9.7 in.)
Unit Size:
Pack of 10
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
10794203 41.6 x 33 cm (16.4 x 13 in.) Pack of 10
11777968 41.8 x 24.7 cm (16.5 x 9.7 in.) Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10794203 Supplier Kimberly-Clark™ Supplier No. 7568
Only null left
Add to basket
Add to basket
Extended use cleaning cloths designed to support colour-coded cleaning. Red cleaning cloths are designed for light and medium duty wiping in food processing environments and healthcare environments. Food contact certified.
  • Each pack contains red cleaning cloths
  • 1 ply, durable wiping cloths are designed to support colour-coded cleaning
  • Food contact certified, available in 4 different colours to help avoid cross-contamination
  • Advanced Hydroknit™ technology ensures each cloth is strong and fast absorbing
  • Designed to enhance regulatory workplace hygiene practices and compliance
  • Specially designed for light and medium duty wiping tasks and particularly suited to food processing and healthcare environments
  • Available in a hygienic poly pack helping provide protection from contaminants
  • Single sheet dispensing helps to prevent grime/dirt touching the cleaning wipes while also controlling usage
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleaning Cloth
Length (English) 9.055 in.
Length (Metric) 36.3 cm
Width (English) 9.055 in
Width (Metric) 23.4 cm
Material Paper Fiber, Polypropylene
Color Red
Dimensions (L x W) 41.6 x 33 cm (16.4 x 13 in.)
Format Interfold
For Use With (Application) For industrial areas where heavy-duty task segregation is required
Packaging Quantity 80 Pk.
Ply 1-ply