Learn More
Thermo Scientific™ Remel™ MacConkey Agar
Description
Thermo Scientific™ Remel MacConkey Agar is used for selective and differential isolation of gram-negative bacilli on the basis of lactose fermentation.
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: BAM, COMPF, SMWW, USP. |
| Description | MacConkey Agar |
| Form | Powder |
| Product Type | Agar |
| Quantity | 500 g |