Learn More
- Home
- Products
- Specialty Lab Equipment, Instruments, and Apparatuses
- Mills
- RETSCH ZM200 Ultra Centrifugal Mill: Ring Sieves
RETSCH ZM200 Ultra Centrifugal Mill: Ring Sieves
Product Code. 10019060 Shop All Retsch Products
Change view
Click to view available options
Material:
Stainless Steel
Wear-resistant Stainless Steel
Pore Size:
0.003 in. (0.08 mm)
0.01 in. (0.50 mm)
0.029 in. (0.75 mm)
0.03 in. (1.0 mm)
0.07 in. (2.0 mm)
0.08 mm
0.11 in. (3.0 mm)
0.12 mm
0.20 mm
0.23 in. (6.0 mm)
0.25 mm
0.75 mm
1.0 mm
1.50 mm
2.0 mm
2.00 mm
3.0 mm
3.00 mm
4.0 mm
5.0 mm
5.00 mm
6.0 mm
Pore Style:
Conidur holes
Round holes
Trapezoid holes
Product Type:
Ring Sieve
Ring Sieve with Reinforced Rim
Unit Size:
Each
32 product options available for selection
| Product Code. | Material | Pore Size | Pore Style | Product Type | unitSize |
|---|---|---|---|---|---|
| 10019060 | Stainless Steel | 1.50 mm | Trapezoid holes | Ring Sieve | Each |
| 10498722 | Stainless Steel | 0.01 in. (0.50 mm) | Conidur holes | Ring Sieve | Each |
| 11944242 | Stainless Steel | 1.50 mm | Trapezoid holes | Ring Sieve with Reinforced Rim | Each |
| 10354721 | Stainless Steel | 1.0 mm | Trapezoid holes | Ring Sieve | Each |
| 10519141 | Stainless Steel | 0.12 mm | Trapezoid holes | Ring Sieve | Each |
| 10670252 | Stainless Steel | 0.20 mm | Trapezoid holes | Ring Sieve | Each |
| 10350061 | Stainless Steel | 0.08 mm | Trapezoid holes | Ring Sieve | Each |
| 10655271 | Stainless Steel | 0.75 mm | Trapezoid holes | Ring Sieve | Each |
| 10130831 | Stainless Steel | 2.00 mm | Trapezoid holes | Ring Sieve | Each |
| 11737704 | Stainless Steel | 4.0 mm | Round holes | Ring Sieve | Each |
| 10079340 | Stainless Steel | 0.25 mm | Trapezoid holes | Ring Sieve | Each |
| 10730661 | Stainless Steel | 3.00 mm | Round holes | Ring Sieve | Each |
| 10039070 | Stainless Steel | 5.0 mm | Round holes | Ring Sieve | Each |
| 11761846 | Stainless Steel | 0.029 in. (0.75 mm) | Conidur holes | Ring Sieve with Reinforced Rim | Each |
| 11904242 | Stainless Steel | 0.003 in. (0.08 mm) | Conidur holes | Ring Sieve with Reinforced Rim | Each |
| 10297790 | Stainless Steel | 6.0 mm | Round holes | Ring Sieve | Each |
| 11914242 | Stainless Steel | 0.12 mm | Trapezoid holes | Ring Sieve with Reinforced Rim | Each |
| 11924242 | Stainless Steel | 0.20 mm | Trapezoid holes | Ring Sieve with Reinforced Rim | Each |
| 11954242 | Stainless Steel | 0.07 in. (2.0 mm) | Conidur holes | Ring Sieve with Reinforced Rim | Each |
| 10101543 | Stainless Steel | 0.03 in. (1.0 mm) | Conidur holes | Ring Sieve with Reinforced Rim | Each |
| 11974242 | Stainless Steel | 0.11 in. (3.0 mm) | Round holes | Ring Sieve with Reinforced Rim | Each |
| 11964242 | Stainless Steel | 4.0 mm | Round holes | Ring Sieve with Reinforced Rim | Each |
| 11994242 | Stainless Steel | 0.23 in. (6.0 mm) | Round holes | Ring Sieve with Reinforced Rim | Each |
| 11984242 | Stainless Steel | 5.00 mm | Round holes | Ring Sieve with Reinforced Rim | Each |
| 10656320 | Stainless Steel | 0.01 in. (0.50 mm) | Conidur holes | Ring Sieve with Reinforced Rim | Each |
| 11944252 | Wear-resistant Stainless Steel | 0.25 mm | Trapezoid holes | Ring Sieve with Reinforced Rim | Each |
| 11702513 | Wear-resistant Stainless Steel | 0.20 mm | Trapezoid holes | Ring Sieve with Reinforced Rim | Each |
| 11954252 | Wear-resistant Stainless Steel | 0.75 mm | Trapezoid holes | Ring Sieve with Reinforced Rim | Each |
| 11984252 | Wear-resistant Stainless Steel | 2.0 mm | Trapezoid holes | Ring Sieve with Reinforced Rim | Each |
| 11846013 | Wear-resistant Stainless Steel | 1.0 mm | Trapezoid holes | Ring Sieve with Reinforced Rim | Each |
| 11994252 | Wear-resistant Stainless Steel | 3.0 mm | Round holes | Ring Sieve with Reinforced Rim | Each |
| 11974252 | Wear-resistant Stainless Steel | 1.50 mm | Trapezoid holes | Ring Sieve with Reinforced Rim | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 32 options available.
Este artículo no se puede devolver.
Vea la política de devoluciones
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
Especificaciones
| Product Type | Ring Sieve |
| For Use With (Equipment) | ZM200 ultra centrifugal mil |
| Material | Stainless Steel |
| Pore Size | 1.50 mm |
| Pore Style | Trapezoid holes |
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?