Learn More
- Home
- Products
- Centrifuges
- General Centrifuge Accessories
- Eppendorf™ Rotor Lids
Eppendorf™ Rotor Lids
Product Code. 13246958 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
A-2-MTP
Centrifuge 5427R, FA-45-24-11-Kit
Centrifuge 5427R, FA-45-30-11
F-34-6-38
F-45-17-17-Cyro
F-45-30-11
F-45-48-11
F-45-64-5-PCR
FA-45-16-17
FA-45-24-11
FA-45-24-11-HS
FA-45-24-11-Kit
FA-45-30-11
FA-45-48-11
S-24-11-AT
Material:
Aluminum
Aluminum, Coated
Polypropylene
Description:
Aerosol-tight
Aerosol-tight, QuickLock
Coated
Non-aerosol-tight
Wind shield upper shell
Unit Size:
Each
22 product options available for selection
| Product Code. | For Use With (Equipment) | Material | Description | unitSize |
|---|---|---|---|---|
| 13246958 | FA-45-48-11 | Aluminum | Aerosol-tight, QuickLock | Each |
| 12334948 | A-2-MTP | Aluminum | Wind shield upper shell | Each |
| 15234536 | Centrifuge 5427R, FA-45-24-11-Kit | Aluminum | Aerosol-tight | Each |
| 10791755 | F-34-6-38 | Aluminum | Non-aerosol-tight | Each |
| 15244536 | Centrifuge 5427R, FA-45-30-11 | Aluminum | Aerosol-tight | Each |
| 10197523 | F-45-17-17-Cyro | Polypropylene | Non-aerosol-tight | Each |
| 15254536 | F-45-30-11 | Polypropylene | Non-aerosol-tight | Each |
| 10005693 | F-45-30-11 | Polypropylene | Coated | Each |
| 10782135 | F-45-64-5-PCR | Aluminum | Non-aerosol-tight | Each |
| 15224536 | FA-45-24-11 | Aluminum | Aerosol-tight | Each |
| 13226589 | FA-45-16-17 | Aluminum | Aerosol-tight | Each |
| 10279323 | FA-45-24-11-HS | Aluminum | Aerosol-tight | Each |
| 13246938 | F-45-48-11 | Polypropylene | Non-aerosol-tight | Each |
| 10269753 | FA-45-24-11-Kit | Aluminum | Aerosol-tight | Each |
| 13286948 | FA-45-24-11-Kit | Aluminum | Aerosol-tight, QuickLock | Each |
| 10072383 | FA-45-30-11 | Aluminum | Aerosol-tight | Each |
| 10496132 | FA-45-30-11 | Aluminum | Non-aerosol-tight | Each |
| 13216958 | FA-45-30-11 | Aluminum | Aerosol-tight, QuickLock | Each |
| 10076213 | FA-45-30-11 | Aluminum, Coated | Aerosol-tight | Each |
| 15274536 | FA-45-48-11 | Aluminum | Aerosol-tight | Each |
| 15284536 | FA-45-48-11 | Polypropylene | Non-aerosol-tight | Each |
| 13236948 | S-24-11-AT | Aluminum | Aerosol-tight | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Replacement rotor lids
- Aerosol-tight and aluminum lids available
- Available in a variety of sizes
Specifications
Specifications
| Type | Rotor Lid |
| Material | Aluminum |
| For Use With (Equipment) | FA-45-48-11 |
| Description | Aerosol-tight, QuickLock |