Learn More
Sartorius Vivaspin™ 20 Centrifugal Concentrator
BIOPHARMA
Product Code. p-4187757 Shop All Sartorius Products
Change view
Click to view available options
MWCO:
0.2 μm
10 kDa
100 kDa
1000 kDa
3 kDa
30 kDa
300 kDa
5 kDa
50 kDa
Starter Pack 2 each of: 5K, 10K, 30K, 50K, 100K Da
Quantity:
12/Pk.
48/Pk.
Unit Size:
Pack of 12
Pack of 48
Pore Size:
0.2 μm
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO Quantity unitSize Pore Size
10757651 100 kDa 48/Pk. Pack of 48 -
10365171 1000 kDa 48/Pk. Pack of 48 -
10506282 30 kDa 12/Pk. Pack of 12 -
10623002 Starter Pack 2 each of: 5K, 10K, 30K, 50K, 100K Da 12/Pk. Pack of 12 -
10203671 0.2 μm 48/Pk. Pack of 48 0.2 μm
10385121 30 kDa 48/Pk. Pack of 48 -
10099490 50 kDa 48/Pk. Pack of 48 -
10672422 100 kDa 12/Pk. Pack of 12 -
16656762 10 kDa 48/Pk. Pack of 48 -
10738231 10 kDa 48/Pk. Pack of 48 -
10039460 1000 kDa 12/Pk. Pack of 12 -
10315271 5 kDa 48/Pk. Pack of 48 -
10096881 300 kDa 48/Pk. Pack of 48 -
10273521 50 kDa 12/Pk. Pack of 12 -
16636762 10 kDa 12/Pk. Pack of 12 -
10670333 10 kDa 12/Pk. Pack of 12 -
10012561 3 kDa 12/Pk. Pack of 12 -
10325271 5 kDa 12/Pk. Pack of 12 -
10411052 300 kDa 12/Pk. Pack of 12 -
10652232 0.2 μm 12/Pk. Pack of 12 0.2 μm
10638842 3 kDa 48/Pk. Pack of 48 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
Product Code. 10757651 Supplier Sartorius Supplier No. VS2042
Offers increased volume flexibility and performance. VivaspinTM 20 Centrifugal Concentrator handles up to 20mL in swing bucket rotors and 14mL in fixed angle rotors accepting 50mL centrifuge tubes. Features twin vertical membranes for unparalleled filtration speed and >100-fold concentration.
Offers increased volume flexibility and performance. VivaspinTM 20 Centrifugal Concentrator handles up to 20mL in swing bucket rotors and 14mL in fixed angle rotors accepting 50mL centrifuge tubes. Features twin vertical membranes for unparalleled filtration speed and >100-fold concentration.
Vivaspin™ 20 centrifugal concentrators have been developed to offer increased volume flexibility and performance. Vivaspin™ 20 handles up to 20mL in swing bucket rotors and 14mL in fixed angle rotors accepting 50mL centrifuge tubes. Vivaspin™ 20 features twin vertical membranes for unparalleled filtration speed and >100-fold concentration. Volume graduation allows to monitor the concentration process and the modified dead stop pocket simplifies pipetting of the final concentrate. For higher process flexibility, Vivaspin™ 20 is available with unique accessories and operating methods. For one-step desalting following concentration, a diafiltration cup filled with buffer is placed on the concentrator and spun to achieve 98% salt removal, instead of two spins to achieve the same result with the traditional refill and re-spin procedure. When an appropriate centrifuge is unavailable, or for single sample processing, for gas pressure filtration, Vivaspin™ 20 can be filled with up to 15ml and then pressurised for bench top concentration. For even faster processing, gas pressure filtration can be combined with centrifugation. Pressure-fugation is particularly useful for difficult or viscous samples such as serum, when using a low process temperature, or generally when minimum process time is essential.
  • High flux PES membrane
  • High retentate recovery > 90 %
  • Vertical twin membrane for ultra fast concentration
  • Universal rotor compatibility
  • Safe sample concentration due to dead-stop volume
  • Convenient concentrate recovery by pipetting
  • Volume graduation for easy reading/monitoring
  • 100-fold concentration with twin vertical membranes
  • Additional option for pressurized operation
Rotor Compatibility
Please note: Vivaspin™ 20 (30mm x 116mm) is designed to fit into rotors that can accommodate Falcon 50 ml conical bottom tubes, e.g. Beckman Allegra 25R with TS-5.1-500 swing-out rotor with BUC 5 buckets and 368327 adaptors; Beckman TA-10.250 25° fixed angle rotor with 356966 adaptors; Multifuge 3 S-R with 75006445 swing out rotor with 75006441 buckets and adaptors for Falcon 50 ml conical bottom tubes. These devices are not designed to fit into rotors that only accept round bottom 29mm x 105mm tubes, e.g. Sorvall SS34 or Beckmann JA 20. If your rotor accepts only 29mm x 105mm round bottom tubes, please use the Vivaspin™ 15, which can be used in either round bottom or conical centrifuge tubes
TRUSTED_SUSTAINABILITY
Specifications
Disposable Single-use
Filter Type Ultrafiltration
For Use With (Equipment) Centrifuge
Material (Membrane) PES
MWCO 100 kDa
Sample Volume 20 mL
Sterility Non-sterile
Quantity 48/Pk.
Type Centrifugal Concentrator
Volume (Metric) 14 mL
Length (Metric) 116 mm