missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIGLEC15, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA548221
Additional Details : Weight : 0.01000kg
Specifications
| SIGLEC15 | |
| Polyclonal | |
| PBS with 0.09% sodium azide | |
| Q6ZMC9 | |
| Sialic acid-binding Ig-like lectin 15, Siglec-15, CD33 antigen-like 3, SIGLEC15, CD33L3 | |
| Rabbit | |
| IgG | |
| 400μL | |
| Store at 2° to 8°C for up to 6 months. For long term storage store at -20°C. | |
| Primary | |
| 284266 |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| SIGLEC15 | |
| Liquid | |
| SIGLEC15 | |
| KLH conjugated synthetic peptide between 274-302 amino acids from the C-terminal region of human SIGLEC15. | |
| Antigen affinity chromatography | |
| RUO | |
| Antibody | |
| Polyclonal | |
| Human |