Learn More
RETSCH Sintered Aluminum Oxide Grinding Balls
Product Code. 11954382 Shop All Retsch Products
Change view
Click to view available options
Diameter (Metric):
15 mm
20 mm
30 mm
40 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
11954382 15 mm Each
10350522 40 mm Each
10438161 30 mm Each
10574112 20 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11954382 Supplier RETSCH Supplier No. 05.368.0112
Specifications
Product Type Grinding Balls
Diameter (Metric) 15 mm
For Use With (Equipment) PM 100, PM 200, PM 400 Planetary Ball Mills
Material Sintered Aluminum Oxide