Learn More
Eppendorf™ Spare Sealing for Caps, S-4x400 Rotor
Product Code. 15893222 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
S-4x400 Rotor 400 mL bucket caps
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15893222 S-4x400 Rotor 400 mL bucket caps Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15893222 Supplier Eppendorf™ Supplier No. 5910710003
Only null left
Add to basket
Add to basket
Spare seals for aerosol-tight caps help secure bucket contents when working with hazardous materials.
  • Suitable for use with S-4x400 Rotor bucket caps
  • Helps maintain aerosol-tight seal when working with hazardous substances
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) S-4x400 Rotor 400 mL bucket caps