Learn More
RETSCH Stainless Steel Cutting Container
Use these containers with GRINDOMIX GM 300 Knife Mills. Retsch™ Stainless Steel Cutting Containers are ideal for homogenizing substances with a high water, oil or fat content as well as for grinding dry, soft and medium-hard products.
Brand:  RETSCH 02.045.0055
Additional Details : Weight : 2.05000kg
Product Code. 10490305
  • 976.14€ / Each
please call +351214253350
Add to basket
Description
Description
  • Process sample volumes up to 4.5 liters quickly and reproducibly
  • Ideal for homogenizing foodstuffs
  • Includes knife holder
Specifications
Specifications
Stainless Steel
For Mill GRINDOMIX GM 300
Cutting Container