Learn More
Nickel Electro™ Stainless Steel Beakers with Spout
Product Code. p-8013895 Shop All Nickel-Electro Products
Change view
Click to view available options
Capacity (Metric):
200 mL
350 mL
550 mL
700 mL
1000 mL
1250 mL
2000 mL
2500 mL
5000 mL
Unit Size:
Each
Pack of 5
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11756567 200 mL Each
12737856 350 mL Each
12747856 550 mL Each
11766617 550 mL Pack of 5
11419569 700 mL Each
11429569 1000 mL Each
11439569 1250 mL Each
11449569 2000 mL Each
11459569 2500 mL Each
11469569 5000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 11756567 Supplier Nickel Electro™ Supplier No. 201
Only null left
Add to basket
Edge
Add to basket
Will not shatter or chip, this durable stainless steel laboratory beaker is suitabale for laboratory applications.
  • Durable stainless steel laboratory beaker
  • 304 spec stainless steel beakers 
  • Will not shatter or chip
  • Suitable for laboratory applications since 
  • All surfaces are non-reacting 
  • Offer zero absorption
Specifications
Diameter (Metric) Outer 63 mm
Capacity (Metric) 200 mL
Material Stainless Steel
Height (Metric) 73 mm
Includes Spout
Type Beaker