Learn More
Techne™ Aluminum Dri-Block Insert
Ensure efficient heat transfer with anodized aluminum alloy blocks. Aluminum Techne™ Dri-Block Inserts provide excellent thermal conductivity.
174.00€ - 273.00€
| Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
|
12071602
|
Techne™
F4464 |
1.5mL tubes |
174.00€
Each Qty Price Break Available |
05-01-2021 |
12952590
|
Techne™
F4501 |
12mm dia. Tubes |
273.00€
Each Qty Price Break Available |
25-02-2021 |
11880705
|
Techne™
F3503 |
10mm dia. Tubes |
174.00€
Each Qty Price Break Available |
12-01-2021 |
Description
- For use with Techne™ Dri-Block™ heaters
- Most blocks have a separate hole to accommodate a thermometer if required.
- Techne™ Dri-Block™ heaters can accommodate block inserts in any combination (size-permitting)
- Depending on the combination of blocks selected, plastic spacers can be employed to fill a half-width recess, ensuring optimal thermal performance
- A close fitting of sample tubes ensures efficient heat transfer
- Exceptionally high thermal temperature control uniformity
- Half width blocks have dimensions (D x W x H) 95 x 37 x 50 mm
- Single width blocks have dimensions (D x W x H) 95 x 75 x 50 mm
- Double width blocks have dimensions (D x W x H) 95 x 150 x 61 mm
- Triple width blocks have dimensions (D x W x H) 95 x 225 x 50 mm