missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Techne™ High Temperature Anodized Aluminum Dri-Block™ Insert
Precision-engineered aluminum heating blocks offer excellent temperature accuracy
Brand: Techne™ F4467
Additional Details : Weight : 0.50000kg
Description
- Versatile range of interchangeable heating blocks work with many size sample tubes and plates
- Heaters sold separately
Specifications
| 95 mm | |
| 61 mm | |
| 150 mm |
| Anodized Aluminum | |
| With thermometer hole | |
| Accessory, Block |