missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios Surfa'Safe Premium Foaming Spray

Product Code. 15318954
Change view
Click to view available options
Volume (Metric):
750 mL
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
15318954 750 mL Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15318954 Supplier Laboratoires Anios Supplier No. 2419544

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Disinfecting detergent foam for cleaning and disinfection of surfaces, external equipment structures and non-submersible and non-invasive medical devices.

Detergent disinfectant foam for surfaces and medical devices

  • Cleans and disinfects in a single operation
  • Without CMR, without alcool, without perfume nor colorant
  • Broad spectrum antimicrobial and optimised contact time
  • Compact foam dispenser to limit the formation of aerosols and
    volatile components (750ml)
  • Formula ensures compatibility with a wide range of materials,
    including polymers (e.g.: polycarbonate, plexiglas...)
  • Dense foam for better surface adhesion (750ml)
  • Surfaces dry fast without marks
  • Type 4 biocide which can be used on surfaces which come into
    contact with food products, followed by rinsing
  • Improved ecotoxicological profile: GHG*
    emissions reduced by 7% relative to a similar product
  • pH : 6 ± 0.5

Specifications

Product Type Foaming Disinfectant
For Use With (Application) For Cleaning and Disinfection of Surfaces, External Structures of Equipment
Quantity 12/Pk.
Volume (Metric) 750 mL
Product Identifier
  • SURFA'SAFE PREMIUM
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: DIDECYLDIMETHYLAMMONIUM CHLORIDE, N-C12-14-ALKYLTRIMETHYLENEDI-AMINES
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.