Learn More
Nabertherm Tabletop Forced Convection Chamber Furnace
Equipped for soft annealing of copper or tempering of titanium, and also for annealing of steel under non-flammable protective or reaction gases. The modular forced convection chamber furnace design allows for adaptation to specific process requirements with appropriate accessories.
Brand:  Nabertherm 001334150
Additional Details : Weight : 50.00000kg
Product Code. 11947445
  • 4659.77€ / Each
please call +351214253350
Add to basket
Description
Description
  • All parts may be ordered separately
  • The hardening system can be retrofitted or equipment added individually depending on the materials processed
  • The MHS 17 can have an air quenching system added to it for air-hardened steels.
  • This platform has a powerful cooling fan to force cool the parts requiring hardening and also the gas feed annealing bag and holder.
  • A refractory brick base is for placing hot boxes and workpieces on them.
  • The quenching baths can also be fastened onto the forced cooling system.
  • An additional storage platform can be integrated within the system for holding accessory equipment and/or optional charging accessories
Specifications
Specifications
340 x 295 x 170 mm
110 - 240 V, 50/60Hz
55 kg
Up to 650°C
845 x 470 x 460 mm
P330
15 L